Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035309.1 | NP_001030386.1 | 4 - 5 | 497 | 77 | |
GenBank mRNA | BC105228.1 | - | 4 - 5 | 497 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |