Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040585.2 | NP_001035675.1 | 1 - 2 | 161 | 105 | |
GenBank mRNA | BC109923.1 | - | 1 - 2 | 162 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |