Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038033.2 | NP_001033122.1 | 2 - 3 | 245 | 103 | |
GenBank mRNA | BC111240.1 | - | 2 - 3 | 246 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |