Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038163.2 | NP_001033252.1 | 3 - 4 | 452 | 99 | |
GenBank mRNA | BC110002.1 | - | 3 - 4 | 455 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |