Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039962.1 | NP_001035051.1 | 1 - 2 | 106 | 81 | |
GenBank mRNA | DQ319072.1 | - | 1 - 2 | 106 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |