Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045913.1 | NP_001039378.1 | 4 - 5 | 779 | 98 | |
GenBank mRNA | BC112722.1 | - | 4 - 5 | 779 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |