Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077072.1 | NP_001070540.1 | 1 - 2 | 326 | 104 | |
GenBank mRNA | BC111230.1 | - | 1 - 2 | 326 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |