Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079798.1 | NP_001073266.1 | 5 - 6 | 880 | 65 | |
GenBank mRNA | BC109692.1 | - | 5 - 6 | 880 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |