Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046359.1 | NP_001039824.1 | 1 - 2 | 333 | 74 | |
GenBank mRNA | BC114130.1 | - | 1 - 2 | 333 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |