Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046474.2 | NP_001039939.1 | 3 - 4 | 767 | 55 | |
GenBank mRNA | AJ871967.1 | - | 3 - 4 | 741 | 55 | |
AJ871968.1 | - | 3 - 4 | 597 | 55 | ||
BC118079.1 | - | 3 - 4 | 768 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |