Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22H3orf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046510.1 | NP_001039975.1 | 3 - 4 | 953 | 82 | |
GenBank mRNA | BC113216.1 | - | 3 - 4 | 953 | 82 | |
DQ335871.1 | - | 2 - 3 | 191 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |