Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075127.2 | NP_001068595.1 | 2 - 3 | 414 | 63 | |
GenBank mRNA | BC118429.1 | - | 2 - 3 | 415 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |