Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075518.1 | NP_001068986.1 | 7 - 8 | 1592 | 87 | |
GenBank mRNA | BC147958.1 | - | 8 - 9 | 1138 | 87 | |
BT025453.1 | - | 7 - 8 | 1592 | 87 | ||
BT026328.1 | - | 8 - 9 | 1392 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |