Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075552.1 | NP_001069020.1 | 2 - 3 | 291 | 101 | |
GenBank mRNA | BC114715.1 | - | 2 - 3 | 291 | 101 | |
BT025449.1 | - | 2 - 3 | 308 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |