Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076458.2 | NP_001069926.1 | 2 - 3 | 281 | 66 | |
GenBank mRNA | BC116155.1 | - | 2 - 3 | 282 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |