Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076478.1 | NP_001069946.1 | 1 - 2 | 254 | 116 | |
GenBank mRNA | BC104535.1 | - | 1 - 2 | 222 | 116 | |
BT025365.1 | - | 1 - 2 | 254 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |