Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075795.2 | NP_001069263.1 | 1 - 2 | 71 | 57 | |
NM_001206416.1 | NP_001193345.1 | 1 | 71 | 57 | ||
GenBank mRNA | BC122831.1 | - | 1 - 2 | 72 | 57 | |
EH132414.1 | - | 1 - 2 | 72 | 57 | ||
EV682356.1 | - | 1 - 2 | 54 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |