Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075795.2 | NP_001069263.1 | 5 - 6 | 576 | 125 | |
NM_001206416.1 | NP_001193345.1 | 1 | 576 | 125 | ||
GenBank mRNA | BC122831.1 | - | 5 - 6 | 577 | 125 | |
EH132414.1 | - | 5 - 6 | 577 | 125 | ||
EV682356.1 | - | 5 - 6 | 559 | 125 | ||
EV746051.1 | - | 1 | 430 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |