Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076011.1 | NP_001069479.1 | 5 - 6 | 1764 | 76 | |
GenBank mRNA | BC119830.1 | - | 5 - 6 | 1764 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |