Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098913.2 | NP_001092383.1 | 4 - 5 | 490 | 145 | |
GenBank mRNA | BC146109.1 | - | 4 - 5 | 491 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |