Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098929.1 | NP_001092399.1 | 5 - 6 | 481 | 124 | |
GenBank mRNA | BC146185.1 | - | 5 - 6 | 481 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |