Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102088.2 | NP_001095558.1 | 4 - 5 | 518 | 149 | |
GenBank mRNA | BC151501.1 | - | 4 - 5 | 519 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |