Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYROXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105334.1 | NP_001098804.1 | 6 - 7 | 651 | 142 | |
GenBank mRNA | BC123726.1 | - | 6 - 7 | 651 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |