Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 9 - 10 | 788 | 91 | |
GenBank mRNA | BC105446.1 | - | 9 - 10 | 824 | 91 | |
DV883546.1 | - | 6 - 7 | 470 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |