Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076426.2 | NP_001069894.2 | 5 - 6 | 614 | 82 | |
GenBank mRNA | BC102597.1 | - | 5 - 6 | 614 | 81 | |
DT841167.1 | - | 5 - 6 | 614 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |