Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076455.2 | NP_001069923.2 | 3 - 4 | 240 | 73 | |
GenBank mRNA | BC109857.1 | - | 3 - 4 | 230 | 73 | |
DT834350.1 | - | 3 - 4 | 241 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |