Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101176.1 | NP_001094646.1 | 3 - 4 | 485 | 64 | |
GenBank mRNA | BC151644.1 | - | 3 - 4 | 485 | 64 | |
BT025430.1 | - | 3 - 4 | 613 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |