Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102188.1 | NP_001095658.1 | 5 - 6 | 768 | 79 | |
GenBank mRNA | BC149178.1 | - | 5 - 6 | 768 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |