Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110446.2 | NP_001103916.1 | 2 | 150 | 140 | |
GenBank mRNA | BC109623.1 | - | 2 | 152 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Amplicon spans exons and probe does not span exons |