Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSIG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205873.1 | NP_001192802.1 | 2 - 3 | 360 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |