Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206350.2 | NP_001193279.1 | 5 - 6 | 833 | 87 | |
GenBank mRNA | BC149130.1 | - | 5 - 6 | 833 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |