Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR3LG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206792.1 | NP_001193721.1 | 4 - 5 | 1116 | 114 | |
GenBank mRNA | EH166075.1 | - | 3 - 4 | 517 | 114 | |
MH237865.1 | - | 4 - 5 | 1020 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |