Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193009.2 | NP_001179938.1 | 6 - 7 | 930 | 60 | |
GenBank mRNA | EH210414.1 | - | 3 - 4 | 396 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |