Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193253.1 | NP_001180182.1 | 3 - 4 | 1414 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |