Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199762.1 | NP_001186691.1 | 1 - 2 | 401 | 71 | |
GenBank mRNA | BC102715.1 | - | 1 - 2 | 265 | 71 | |
DT807444.1 | - | 1 - 2 | 184 | 71 | ||
DT829639.1 | - | 1 - 2 | 265 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |