Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078091.5 | NP_510492.2 | 5 - 6 | 634 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() |