Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306444.3 | NP_001293373.1 | 11 - 12 | 3067 | 66 | |
NM_001306445.3 | NP_001293374.1 | 6 - 7 | 2082 | 66 | ||
NM_001306446.3 | NP_001293375.1 | 6 - 7 | 2100 | 66 | ||
NM_001377813.3 | NP_001364767.1 | 12 - 13 | 2754 | 66 | ||
NM_001377816.2 | NP_001364504.1 | 12 - 13 | 2776 | 66 | ||
NM_170837.5 | NP_740821.1 | 11 - 12 | 3052 | 66 | ||
GenBank mRNA | AF308860.1 | - | 11 - 12 | 3051 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |