Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062999.9 | NP_495400.2 | 12 - 13 | 2474 | 100 | |
NM_171014.6 | NP_741015.1 | 12 - 13 | 2471 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |