Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exos-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372617.1 | NP_001360028.1 | 4 - 5 | 368 | 90 | |
NM_171186.6 | NP_741217.1 | 5 - 6 | 390 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |