Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171383.5 | NP_741446.1 | 1 - 2 | 54 | 68 | |
NR_131573.1 | 1 - 2 | 53 | 68 | |||
NR_131574.1 | 1 - 2 | 53 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |