Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asic-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059698.3 | NP_492099.1 | 11 - 12 | 1435 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |