Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rskn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026431.3 | NP_001021602.1 | 8 - 9 | 1571 | 82 | |
NM_001026432.4 | NP_001021603.1 | 9 - 10 | 1584 | 82 | ||
NM_001129032.4 | NP_001122504.1 | 9 - 10 | 1590 | 82 | ||
NM_001373180.3 | NP_001359590.1 | 10 - 11 | 1806 | 82 | ||
NM_059918.4 | NP_492319.2 | 9 - 10 | 1634 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |