Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059944.8 | NP_492345.2 | 5 - 6 | 546 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |