Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026296.3 | NP_001021467.1 | 4 - 5 | 1912 | 89 | |
NM_001026297.6 | NP_001021468.1 | 4 - 5 | 787 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |