Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026488.4 | NP_001021659.1 | 3 - 4 | 606 | 73 | |
NM_001026489.7 | NP_001021660.2 | 16 - 17 | 3572 | 73 | ||
NM_001026490.6 | NP_001021661.2 | 16 - 17 | 3570 | 73 | ||
NM_001306400.4 | NP_001293329.1 | 16 - 17 | 3574 | 73 | ||
NM_001306401.3 | NP_001293330.1 | 11 - 12 | 2709 | 73 | ||
NM_001306402.3 | NP_001293331.1 | 11 - 12 | 2709 | 73 | ||
NM_001306403.3 | NP_001293332.1 | 10 - 11 | 2523 | 73 | ||
NM_001306404.3 | NP_001293333.1 | 10 - 11 | 2523 | 73 | ||
NM_001306405.3 | NP_001293334.1 | 7 - 8 | 2244 | 73 | ||
NM_001306406.3 | NP_001293335.1 | 7 - 8 | 2244 | 73 | ||
NM_001306407.3 | NP_001293336.1 | 5 - 6 | 1998 | 73 | ||
NM_001306408.3 | NP_001293337.1 | 5 - 6 | 1998 | 73 | ||
NM_001306529.4 | NP_001293458.1 | 6 - 7 | 2327 | 73 | ||
NM_001356800.4 | NP_001343732.1 | 13 | 3125 | 73 | ||
NM_058537.5 | NP_490938.3 | 16 - 17 | 3566 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |