Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cps-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058970.5 | NP_491371.1 | 4 - 5 | 579 | 80 | |
GenBank mRNA | AF390558.1 | - | 4 - 5 | 572 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |