Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lam-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060374.5 | NP_492775.2 | 3 - 4 | 966 | 64 | |
GenBank mRNA | AF074902.1 | - | 3 - 4 | 966 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |