Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029819.7 | NP_001024990.1 | 2 - 3 | 516 | 61 | |
NM_171652.7 | NP_741748.2 | 2 - 3 | 514 | 61 | ||
GenBank mRNA | AF041053.1 | - | 2 - 3 | 431 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |