Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171668.9 | NP_741768.1 | 4 - 5 | 620 | 99 | |
NM_171669.6 | NP_741769.1 | 3 - 4 | 437 | 99 | ||
GenBank mRNA | AF386745.1 | - | 3 - 4 | 436 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |