Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375204.1 | NP_001361941.1 | 2 - 3 | 253 | 98 | |
NM_171812.7 | NP_741945.3 | 3 - 4 | 437 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |